missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylbutyrat, 99 %, Thermo Scientific™

Catalog Number p-7025293
Click to view available options
Menge:
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1551-44-6
C10H18O2
170.252
MFCD00046354
VZHUBBUZNIULNM-UHFFFAOYSA-N
cyclohexyl butyrate, butanoic acid, cyclohexyl ester, cyclohexanyl butyrate, butyric acid, cyclohexyl ester, cyclohexyl n-butyrate, unii-fhh53z3i16, fema no. 2351, ccris 6553, cyclohexanol butanoate, cyclohexyl-n-butyrate
243783
Cyclohexylbutanoat
CCCC(=O)OC1CCCCC1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1551-44-6
C10H18O2
170.252
MFCD00046354
VZHUBBUZNIULNM-UHFFFAOYSA-N
cyclohexyl butyrate, butanoic acid, cyclohexyl ester, cyclohexanyl butyrate, butyric acid, cyclohexyl ester, cyclohexyl n-butyrate, unii-fhh53z3i16, fema no. 2351, ccris 6553, cyclohexanol butanoate, cyclohexyl-n-butyrate
243783
Cyclohexylbutanoat
CCCC(=O)OC1CCCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Cyclohexyl butyrate
Dichte 0.94
Siedepunkt 90°C (15 mmHg)
Brechungsindex 1.442
Menge 50 g
Beilstein 2083414
Formelmasse 170.25
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'