missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentansulfonylchlorid, 97 %, Thermo Scientific Chemicals

Catalog Number 15465627
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
26394-17-2
C5H9ClO2S
168.635
MFCD04117774
HZVKYZHPDGEECE-UHFFFAOYSA-N
cyclopentylsulfonyl chloride, cyclopentanesulphonyl chloride, chlorocyclopentylsulfone, cyclopentylsulfonylchloride, acmc-1ccr5, cyclopentanesulfonylchloride, cyclopentyl sulfonyl chloride, chlorosulphonyl cyclopentane, cyclopentane sulfonyl chloride, cyclopentane sulphonyl chloride
3717197
Cyclopentansulfonylchlorid
C1CCC(C1)S(=O)(=O)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
26394-17-2
C5H9ClO2S
168.635
MFCD04117774
HZVKYZHPDGEECE-UHFFFAOYSA-N
cyclopentylsulfonyl chloride, cyclopentanesulphonyl chloride, chlorocyclopentylsulfone, cyclopentylsulfonylchloride, acmc-1ccr5, cyclopentanesulfonylchloride, cyclopentyl sulfonyl chloride, chlorosulphonyl cyclopentane, cyclopentane sulfonyl chloride, cyclopentane sulphonyl chloride
3717197
Cyclopentansulfonylchlorid
C1CCC(C1)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Cyclopentanesulfonyl chloride
Siedepunkt 71°C to 73°C (2 mmHg)
Menge 1 g
UN-Nummer UN3265
Empfindlichkeit Moisture sensitive
Formelmasse 168.64
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'