Learn More
ACROBiosystems Cynomolgus CD73 Protein, His Tag (active enzyme)
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | EHH53214.1 |
---|---|
Molekulargewicht | 60.0 kDa |
Quelle | Cynomolgus CD73, His Tag (CD3-C52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Cys 554 (Accession # EHH53214.1). |
Immunogen | Trp 27 - Cys 554 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17106121
|
ACROBiosystems
CD3-C52H9-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 02-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
17116121
|
ACROBiosystems
CD3-C52H9-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 02-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
5'-nucleotidase (5'-NT), also known as ecto-5'-nucleotidase or CD73 (cluster of differentiation 73), is an enzyme that is encoded by the NT5E gene. CD73 commonly serves to convert AMP to adenosine. Ecto-5-prime-nucleotidase (5-prime-ribonucleotide phosphohydrolase) catalyzes the conversion at neutral pH of purine 5-prime mononucleotides to nucleosides, the preferred substrate being AMP. Other forms of 5-prime nucleotidase exist in the cytoplasm and lysosomes and can be distinguished from ecto-NT5 by their substrate affinities, requirement for divalent magnesium ion, activation by ATP, and inhibition by inorganic phosphate. Rare allelic variants are associated with a syndrome of adult-onset calcification of joints and arteries (CALJA) affecting the iliac, femoral, and tibial arteries reducing circulation in the legs and the joints of the hands and feet causing pain.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.0 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Spezifikation
EHH53214.1 | |
Cynomolgus CD73, His Tag (CD3-C52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Cys 554 (Accession # EHH53214.1). | |
-20°C | |
CD73 | |
Cynomolgus Monkey | |
Tris and NaCl, pH7.5 | |
HEK293 | |
Enzymes |
60.0 kDa | |
Trp 27 - Cys 554 | |
CD73,NT5E,5 '-Nucleotidase,5 '-NT,NT5,NTE | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |