Learn More
ACROBiosystems Cynomolgus LAG-3 / CD223 Protein, Fc Tag
Marke:  ACROBiosystems LA3-C5252-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15887207
  • 2818.49 € / 1 Milligramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Lymphocyte activation gene 3 protein (LAG3) is also known as CD antigen CD223 and protein FDC, which belongs to immunoglobulin (Ig) superfamily and contains 4 extracellular Ig-like domains. The LAG3 gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG3 to CD4. LAG3 /CD223 involved in lymphocyte activation. LAG3 /CD223 binds to HLA class-II antigens.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.0 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
XP_005570011.1
1 mg
Ala 18 - His 449
LAG3,CD223,FDC
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>85% as determined by SDS-PAGE
73.0 kDa
Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_005570011.1).
-20°C
LAG-3
Cynomolgus Monkey
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Immune Checkpoint Proteins