Learn More
Cysteinyl Leukotriene R2/CysLTR2 Antibody, Novus Biologicals™
Artikelnummer. 18484512 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
25 μL
Packungsgröße:
0.10 Milliliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18484512 25 μL 25 Mikroliter
18487781 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18484512 Lieferant Novus Biologicals Lieferanten-Nr. NBP21389725ul
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
Cysteinyl Leukotriene R2/CysLTR2 Polyclonal antibody specifically detects Cysteinyl Leukotriene R2/CysLTR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Cysteinyl Leukotriene R2/CysLTR2
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Zusammensetzung PBS (pH 7.2) and 40% Glycerol
Gen-Alias CysLT(2), CYSLT2KPG_011, CYSLT2RHG57, CysLTR2, cysteinyl leukotriene CysLT2 receptor, cysteinyl leukotriene receptor 2, G protein-coupled receptor, G-protein coupled receptor GPCR21, G-protein coupled receptor HG57, hGPCR21, HPN321
Wirtsspezies Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: KFMSLQPSISVSEMEPNGTFSNNNSRNCTIE
Reinigungsverfahren Immunogen affinity purified
Menge 25 μL
Regulatorischer Status RUO
Forschungsgebiet GPCR
Primär oder sekundär Primary
Gen-ID (Entrez) 57105
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG