missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16687-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
GRP1 (general receptor for phosphoinositides-1) contains a Pleckstrin homology (PH) domain as well as a Sec7 domain. The PH domain has high binding affinity for phosphatidylinositol 3,4,5-trisphosphate Ptdlns(3,4,5)P3), while the Sec7 homology domain is responsible for catalyzing guanine nucleotide exchange of ADP-ribosylation factor (ARF) proteins. GRP1 co-localizes with ARF6 and catalyzes GTP/GDP exchange on ARF6. It is known to interact with Ptdlns(3,4,5)P3 localized to the plasma membrane in vitro and may also be a Ptdlns(3,4,5)P3 receptor. Additionally, GRP1 may regulate protein sorting and membrane trafficking through interaction with the guanosine triphosphate ARF, and may control cell adhesion through interaction with integrins.Spezifikation
Cytohesin 3 | |
Polyclonal | |
Unconjugated | |
CYTH3 | |
ARNO3, CYTH3, cytohesin 3, GRP1, Protein ARNO3, PSCD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116693, 19159, 9265 | |
-20°C | |
Liquid |
Western Blot | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08967, O43739, P97696 | |
Cyth3 | |
Cytohesin 3 Fusion Protein Ag10025 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |