missing translation for 'onlineSavingsMsg'
Learn More

D 4476, Tocris Bioscience™

Selective CK1 inhibitor. Also inhibits TGF-βRI

Marke:  Tocris (Bio-Techne) 290250

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412014

  • 1041.87 € / 50 Milligramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Selective inhibitor of casein kinase 1 (CK1) and TGF-β type-I receptor (ALK5) that displays > 20-fold selectivity over SAPK2/p38 and a much greater selectivity over all other protein kinases tested. Suppresses site-specific phosphorylation and nuclear exclusion of FOXO1a.
Spezifikation

Chemische Identifikatoren

301836-43-1
398.422
ck1 inhibitor, casein kinase i inhibitor, d4476, casein kinase i inhibitor, 4-4-2,3-dihydrobenzo 1,4 dioxin-6-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide, 4-4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide, benzamide,4-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-2-pyridinyl-1h-imidazol-2-yl, 4-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-2-pyridinyl-1h-imidazol-2-yl benzamide, insolution casein kinase i inhibitor, d4476, 4-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide, 4-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide
4-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-pyridin-2-yl-1H-imidazol-2-yl]benzamide
C23H18N4O3
DPDZHVCKYBCJHW-UHFFFAOYSA-N
6419753
C1COC2=C(O1)C=CC(=C2)C3=C(NC(=N3)C4=CC=C(C=C4)C(=O)N)C5=CC=CC=N5

Spezifikation

D 4476
301836-43-1
ck1 inhibitor, casein kinase i inhibitor, d4476, casein kinase i inhibitor, 4-4-2,3-dihydrobenzo 1,4 dioxin-6-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide, 4-4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide, benzamide,4-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-2-pyridinyl-1h-imidazol-2-yl, 4-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-2-pyridinyl-1h-imidazol-2-yl benzamide, insolution casein kinase i inhibitor, d4476, 4-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide, 4-4-2,3-dihydro-1,4-benzodioxin-6-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide
C1COC2=C(O1)C=CC(=C2)C3=C(NC(=N3)C4=CC=C(C=C4)C(=O)N)C5=CC=CC=N5
398.422
398.41
50mg
C23H18N4O3
DPDZHVCKYBCJHW-UHFFFAOYSA-N
4-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-pyridin-2-yl-1H-imidazol-2-yl]benzamide
6419753
>99%
Videos