Learn More
D-AP5, Tocris Bioscience™
Potent and selective NMDA antagonist; more active form of DL-AP5
Marke:  Tocris (Bio-Techne) 0106/10
Weitere Details : Netto-Gewicht : 0.00150kg
Menge. 10mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417433
  • 204.60 € / 10 Milligramm
Voraussichtlicher Liefertermin 01-08-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Widely used competitive NMDA antagonist. More active form of AP5. Also agonist at quisqualate-sensitized AP6 site where it is less potent than the L-isomer, L-AP5 (Cat. No.0107).
Available as part of the Mixed NMDA Receptor Tocriset™ . DL Mixture and L-isomer also available.
Spezifikation
Chemische Identifikatoren
79055-68-8
197.127
d-ap5, 5-phosphono-d-norvaline, d-norvaline, 5-phosphono, d-apv, 2r-2-amino-5-phosphonopentanoic acid, d-2-amino-5-phosphopentanoic acid, d---2-amino-5-phosphonopentanoic acid, d--apv, d-2-amino-5-phosphonovaleric acid, d--ap-5
(2R)-2-amino-5-phosphonopentanoic acid
C5H12NO5P
VOROEQBFPPIACJ-SCSAIBSYSA-N
135342
C(CC(C(=O)O)N)CP(=O)(O)O
Spezifikation
D-AP5
79055-68-8
d-ap5, 5-phosphono-d-norvaline, d-norvaline, 5-phosphono, d-apv, 2r-2-amino-5-phosphonopentanoic acid, d-2-amino-5-phosphopentanoic acid, d---2-amino-5-phosphonopentanoic acid, d--apv, d-2-amino-5-phosphonovaleric acid, d--ap-5
C(CC(C(=O)O)N)CP(=O)(O)O
197.127
197.13
10mg
C5H12NO5P
VOROEQBFPPIACJ-SCSAIBSYSA-N
(2R)-2-amino-5-phosphonopentanoic acid
135342
>99%
Videos