Learn More
D-Galactono-1,4-lacton, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | D-Galactono-1,4-lactone |
| CAS | 2782-07-2 |
| Summenformel | C6 H10 O6 |
| Menge | 250 mg |
| Synonym | D-Galactonic acid, γ-lactone, Galactonic acid, γ-lactone, D- (8CI), 1,4-D-Galactonolactone, D-Galactonic acid 1,4-lactone, D-Galactono-1,4-lactone, D-Galactono-γ-lactone, NSC 102762, γ-D-Galactonolactone |
| InChI-Formel | InChI=1S/C6H10O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-5,7-10H,1H2/t2-,3-,4-,5+/m1/s1 |
| SMILES | OC[C@@H](O)[C@@H]1OC(=O)[C@H](O)[C@H]1O |
| IUPAC-Name | (3R,4R,5S)-5-[(1R)-1,2-DiHydroxyethyl]-3,4-DiHydroxyoxolan-2-on |
| Molekulargewicht (g/mol) | 178.14 |
| Formelmasse | 178.0477 |