missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronamid, 98 %, Thermo Scientific Chemicals

Catalog Number p-7043185
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3789-97-7
C6H11NO6
193.155
MFCD00006619
VOIFKEWOFUNPBN-UHFFFAOYSA-N
glucuronamide, d-glucuronic acid amide, voifkewofunpbn-uhfffaoysa, 3,4,5,6-tetrahydroxytetrahydro-2h-pyran-2-carboxamide non-preferred name
3482
3,4,5,6-Tetrahydroxyoxan-2-Carboxamid
C1(C(C(OC(C1O)O)C(=O)N)O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3789-97-7
C6H11NO6
193.155
MFCD00006619
VOIFKEWOFUNPBN-UHFFFAOYSA-N
glucuronamide, d-glucuronic acid amide, voifkewofunpbn-uhfffaoysa, 3,4,5,6-tetrahydroxytetrahydro-2h-pyran-2-carboxamide non-preferred name
3482
3,4,5,6-Tetrahydroxyoxan-2-Carboxamid
C1(C(C(OC(C1O)O)C(=O)N)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material D-Glucuronamide
Schmelzpunkt 166°C to 167°C
Farbe Braun bis weiß
Menge 1 g
Löslichkeitsinformationen Soluble in water.
Optische Drehung +35° (c=2 in Water)
Formelmasse 193.15
Reinheit (%) 98%
Physikalische Form Pulver

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'