Learn More
Daptomycin, Tocris Bioscience™
Antibiotic; inhibits protein, DNA and RNA synthesis in gram-positive bacteria
Marke:  Tocris (Bio-Techne) 391750
Weitere Details : Netto-Gewicht : 0.01000kg
Menge. 50mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13413814
  • 469.32 € / 50 Milligramm
Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Lipopeptide, calcium-dependent antibiotic. Exhibits potent bacteriocidal activity against most gram-positive bacteria in vitro and in vivo, including antibiotic-resistant strains such as MRSA and VRE. Disrupts plasma membrane function; activity results in membrane depolarization leading to inhibition of protein, DNA and RNA synthesis.
Spezifikation
Chemische Identifikatoren
103060-53-3
1620.693
N-(1-Oxodecyl)-L-tryptophyl-D-asparaginyl-L-α-aspartyl-L-threonylglycyl-L-ornithinyl-L-α-aspartyl-D-alanyl-L-α-aspartylglycyl-D-seryl-(3R)-3-methyl-L-α-glutamyl-α, 2-
CCCCCCCCCC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)NC(CC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC3C(OC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC3=O)CCCN)CC(=O)O)C)CC(=O)O)CO)C(C)CC(=O)O)CC(=O)C4=CC=CC=C4N)C
C72H101N17O26
DOAKLVKFURWEDJ-AVSAZHKKSA-N
133640189
Spezifikation
Daptomycin
C72H101N17O26
DOAKLVKFURWEDJ-AVSAZHKKSA-N
1620.693
1620.67
50mg
103060-53-3
N-(1-Oxodecyl)-L-tryptophyl-D-asparaginyl-L-α-aspartyl-L-threonylglycyl-L-ornithinyl-L-α-aspartyl-D-alanyl-L-α-aspartylglycyl-D-seryl-(3R)-3-methyl-L-α-glutamyl-α, 2-
CCCCCCCCCC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)NC(CC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC3C(OC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC3=O)CCCN)CC(=O)O)C)CC(=O)O)CO)C(C)CC(=O)O)CC(=O)C4=CC=CC=C4N)C
133640189
>95%
Videos