missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
DDX52 Polyclonal antibody specifically detects DDX52 in Human samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | DDX52 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Alexa Fluor 594 |
| Verdünnung | Western Blot |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | ATP-dependent RNA helicase ROK1-like, DEAD (Asp-Glu-Ala-Asp) box polypeptide 52, DEAD box protein 52, EC 3.6.1, HUSSY19, probable ATP-dependent RNA helicase DDX52, ROK1EC 3.6.4.13 |
| Wirtsspezies | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 499 and 549 of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 52 using the numbering given in entry NP_008941.2 (GeneID 11056). |
| Reinigungsverfahren | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?