missing translation for 'onlineSavingsMsg'
Learn More

Decaborane, 98%, Thermo Scientific Chemicals

Product Code. 18874123
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
18874123 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18874123 Supplier Thermo Scientific Alfa Aesar Supplier No. 087892.14
 Restricted Item
Verfügbar ab: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

In rocket propellants, and as a catalyst in olefin polymerization

Decaborane is used as a fuel for aneutronic fusion. It is also used in the preparation of vinyl decaborane as well as in the preparation of semiconductors that have low energy ion implantation. It is widely used in rocket propellants and serves as a catalyst in olefin polymerization reactions. It is a reactant for the preparation of carborane-conjugated quinoline carboxamide ligands of translocator protein for boron neutron capture therapy (BNCT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17702-41-9
Molecular Formula B10H14
Molecular Weight (g/mol) 122.22
MDL Number MFCD00011314
InChI Key KGUZAUWGHAIUGS-UHFFFAOYSA-N
Synonym decaborane, decaboron tetradecahydride
PubChem CID 6328162
IUPAC Name decaborane
SMILES [B][B][B][B][B][B][B][B][B][B]

Specifications

Boiling Point 213°C
Melting Point 99°C to 103°C
Color White through Yellow
Physical Form Powder
Quantity 25 g
UN Number UN1868
Merck Index 14,2845
Solubility Information Soluble in alcohol,toluene,ethyl acetate and benzene; Slightly soluble in cold water.
Formula Weight 122.22
Percent Purity 98%
Odor Pungent
Density 0.940g/mL
Chemical Name or Material Decaborane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.