Learn More
Serva Electrophoresis™ DePeX
Product Code. 11741957
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11741957 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11741957 Supplier Serva Electrophoresis™ Supplier No. 1824301
 Restricted Item
Verfügbar ab: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Mounting Medium
Content And Storage 15° to 30°C
Volume (Metric) 100 mL
compliance-icons compliance-icons
Product Identifier
  • DePeX
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.