Learn More
Deshydroxymethyl-Losartan-Trifluoressigsäuresalz, TRC
Specifications
Specifications
| Chemischer Name oder Material | Deshydroxymethyl Losartan Trifluoroacetic Acid Salt |
| CAS | 1215727-33-5 |
| Summenformel | C21 H21 Cl N6 . C2 H F3 O2 |
| Menge | 5 mg |
| Synonym | Losartan Impurity C |
| InChI-Formel | InChI=1S/C21H21ClN6.C2HF3O2/c1-2-3-8-20-23-19(22)14-28(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)21-24-26-27-25-21;3-2(4,5)1(6)7/h4-7,9-12,14H,2-3,8,13H2,1H3,(H,24,25,26,27);(H,6,7) |
| SMILES | CCCCc1nc(Cl)cn1Cc2ccc(cc2)c3ccccc3c4nn[nH]n4.OC(=O)C(F)(F)F |
| IUPAC-Name | 5-[2-[4-[(2-Butyl-4-chlor-imidazol-1-yl)methyl]phenyl]phenyl]-2H-tetrazol;2,2,2-Trifluoressigsäure |
| Molekulargewicht (g/mol) | 506.91 |
| Formelmasse | 506.1445 |