Learn More
Deuterium Oxide 99.9% D + 0.002% TMSP D4 W/V, Eurisotop
Product Code. 17257456
Encompass
Change view
Click to view available options
Deuterium Oxide; CAS: 7789-20-0; Deuterium (D): 0.002% D; Grade: Research Grade; Color: Colorless; Quantity: 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17257456 Deuterium Oxide; CAS: 7789-20-0; Deuterium (D): 0.002% D; Grade: Research Grade; Color: Colorless; Quantity: 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17257456 Supplier Eurisotop Supplier No. D221H
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Synonym: Heavy Water
Deuterium Oxide
  • Isotopic Enrichment: 99.90% D
  • Quantity: 100 mL
CoA and MSDS available at Eurisotop.com
Specifications
Color Colorless
Deuterium (D) 0.002% D
Quantity 100 mL
CAS 7789-20-0
Melting Point 3.81°C
Molecular Formula H2O
MDL Number MFCD00044636
Synonym Heavy Water
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES [2H]O[2H]
IUPAC Name [(²H)oxy](²H)
Molecular Weight (g/mol) 20.03
Grade Research Grade
Chemical Name or Material Deuterium Oxide