Learn More
Deuterium Oxide 99.9% D + 0.1% TMSP D4, Eurisotop
Product Code. 17247456
Encompass
Change view
Click to view available options
Deuterium Oxide; CAS: 7789-20-0; Deuterium (D): 0.1% D; Grade: Research Grade; Color: Colorless; Quantity: 10 x 0.75 mL; Packaging: Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17247456 Deuterium Oxide; CAS: 7789-20-0; Deuterium (D): 0.1% D; Grade: Research Grade; Color: Colorless; Quantity: 10 x 0.75 mL; Packaging: Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17247456 Supplier Eurisotop Supplier No. D221B
 Restricted Item
Verfügbar ab: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Synonym: Heavy Water
Deuterium Oxide
  • Isotopic Enrichment: 99.90% D
  • Quantity: (10) 0.75 mL
  • Packaging: Ampule
CoA and MSDS available at Eurisotop.com
Specifications
Color Colorless
Deuterium (D) 0.1% D
Packaging Ampule
Quantity 10 x 0.75 mL
CAS 7789-20-0
Melting Point 3.81°C
Molecular Formula H2O
MDL Number MFCD00044636
Synonym Heavy Water
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES [2H]O[2H]
IUPAC Name [(²H)oxy](²H)
Molecular Weight (g/mol) 20.03
Grade Research Grade
Chemical Name or Material Deuterium Oxide