missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexamethason 21- Phosphat-Dinatriumsalz, MP Biomedicals™
Click to view available options
Menge:
1 g
Spezifikation
Chemischer Name oder Material | Dexamethason 21-Phosphat-Dinatriumsalz |
Menge | 1 g |
Formelmasse | 516.4 |
Physikalische Form | Fest |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'