missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
DFNA5 Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot
DFNA5 (deafness, autosomal dominant 5), also known as ICERE-1, is a 496 amino acid protein that is expressed in cochlea tissue, as well as in placenta, brain, heart, liver, lung and pancreas as two alternatively spliced isoforms, designated short and long. Defects in the gene encoding DFNA5 are the cause of non-syndromic sensorineural deafness autosomal dominant type 5 (DFNA5), a form of sensorineural hearing loss that results from damage to one of various structures that receive sound information in the brain. The gene encoding DFNA5 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
Spezifikation
Spezifikation
| Antigen | DFNA5 |
| Anwendungen | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Klon | A1H3 |
| Konzentration | 5.351 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gen | Gsdme |
| Gen-Zugriffsnummer | O60443 |
| Gen-Alias | 2310037D07Rik; 4932441K13Rik; deafness, autosomal dominant 5; deafness, autosomal dominant 5 (human); deafness, autosomal dominant 5 homolog; Dfna5; DFNA5, deafness associated tumor suppressor; Dfna5h; EG14210; Fin15; Gasdermin-E; Gasdermin-E, C-terminal; Gasdermin-E, N-terminal; GSDME; GSDME-CT; GSDME-NT; ICERE1; ICERE-1; Inversely correlated with estrogen receptor expression 1; nonsyndromic hearing impairment protein; non-syndromic hearing impairment protein 5; Non-syndromic hearing impairment protein 5 homolog |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?
