missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15735-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
DHRS2 displays NADPH-dependent dicarbonyl reductase activity in vitro with 3,4-Hexanedione, 2,3-Heptanedione and 1-Phenyl-1,2-propanedione as substrates. DHRS2 do not reductase activity is displayed in vitro with steroids, retinoids and sugars as substrates. This protein may inhibit cell replication.Spezifikation
DHRS2 | |
Polyclonal | |
Unconjugated | |
DHRS2 | |
DHRS2, Dicarbonyl reductase HEP27, HEP27, Protein D, SDR25C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10202 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13268 | |
DHRS2 | |
DHRS2 Fusion Protein Ag8508 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |