missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14560-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
DHRS9 is a 3-alpha-hydroxysteroid dehydrogenase that converts 3-alpha-tetrahydroprogesterone (allopregolone) to dihydroxyprogesterone and 3-alpha-androstanediol to dihydroxyprogesterone.Spezifikation
DHRS9 | |
Polyclonal | |
Unconjugated | |
DHRS9 | |
3 alpha HSD, 3ALPHA HSD, DHRS9, RDH E2, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10170, 170635, 241452 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q58NB6, Q8VD48, Q9BPW9 | |
Dhrs9 | |
DHRS9 Fusion Protein Ag6116 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |