missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylether, +99 %, extra trocken, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-3757577
Click to view available options
Menge:
100 mL
1 L
2.5 L
142-96-1
C8H18O
130.23
MFCD00009461
DURPTKYDGMDSBL-UHFFFAOYSA-N
di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
8909
1-butoxybutane
CCCCOCCCC
142-96-1
C8H18O
130.23
MFCD00009461
DURPTKYDGMDSBL-UHFFFAOYSA-N
di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
8909
1-butoxybutane
CCCCOCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Di-n-butyl ether
Namenshinweis Extra Dry
Schmelzpunkt -95.0°C
Farbe Farblos
Dichte 0.7680g/mL
Siedepunkt 141.0°C
Flammpunkt 25°C
Prozentgehaltsbereich 99% min. (GC)
Verpackung AcroSeal Glasflasche
Strukturformel [CH3(CH2)3]2O
Brechungsindex 1.3978 to 1.3998
Menge 100 mL
Rückstand nach Verdampfung 0.0005% max.
Merck Index 15,1572
Relative Dichte 0.768
Löslichkeitsinformationen Solubility in water: 0.03% (20°C). Other solubilities: miscible with ethanol,ether,acetone
Viskosität 0.69 mPa.s (20°C)
Formelmasse 130.23
Reinheit (%) 99+%
Güte Extra trocken
Physikalische Form Flüssig
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'