Learn More
Diaion™ WK10(H), Ionenaustauschharz, schwach saurer poröser Typ, 2.5 meq/ml auf Polymethacrylat
Pharmazeutika, Dealkalination, Metallgewinnung, Antibiotika, Saccharose-Aufreinigung
Marke: Thermo Scientific Alfa Aesar 046666.A1
Weitere Details : Netto-Gewicht : 1.00000kg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Perlen | |
Weakly acidic porous type, 2.5meq/mL on polymethacrylate | |
1 kg |
Diaion™ WK10 (H), ion exchange resin | |
300 to 1180μm |