missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,e]thiepin-11(6-H)-one, 98 %, Thermo Scientific™

Catalog Number p-7026987
Click to view available options
Menge:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1531-77-7
C14H10OS
226.293
MFCD02093738
JGJDEWXZEIHBNW-UHFFFAOYSA-N
dibenzo b,e thiepin-11 6h-one, 6,11-dihydrodibenzo b,e thiepin-11-one, dibenz b,e thiepin-11 6h-one, 6h-benzo c 1 benzothiepin-11-one, 1-oxo-6,11-dihydrodibenzo b,e thiepin, dothiepinone, 9-thiatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaen-2-one, acmc-1buub
73741
6H-Benzo[c][1]benzothiepin-11-on
C1C2=CC=CC=C2C(=O)C3=CC=CC=C3S1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1531-77-7
C14H10OS
226.293
MFCD02093738
JGJDEWXZEIHBNW-UHFFFAOYSA-N
dibenzo b,e thiepin-11 6h-one, 6,11-dihydrodibenzo b,e thiepin-11-one, dibenz b,e thiepin-11 6h-one, 6h-benzo c 1 benzothiepin-11-one, 1-oxo-6,11-dihydrodibenzo b,e thiepin, dothiepinone, 9-thiatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaen-2-one, acmc-1buub
73741
6H-Benzo[c][1]benzothiepin-11-on
C1C2=CC=CC=C2C(=O)C3=CC=CC=C3S1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Dibenzo[b,e]thiepin-11(6H)-one
Schmelzpunkt 86°C to 88°C
Siedepunkt 175°C to 180°C (1 mmHg)
Menge 5 g
Formelmasse 226.29
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'