missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl 5-Aminoisophthalat, 96 %, Thermo Scientific™

Catalog Number p-7046504
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
152699-63-3
C22H19NO4
361.397
MFCD03093065
VDJWRMQQXQBYIO-UHFFFAOYSA-N
dibenzyl 5-aminoisophthalate, 1,3-dibenzyl 5-aminobenzene-1,3-dicarboxylate, acmc-20alb1, 3,5-dibenzyloxycarbonylaniline, 3,5-dibenzyloxycarbonyl-aniline, 5-amino-isophthalic acid dibenzyl ester, phenylmethyl 5-amino-3-benzyloxycarbonyl benzoate, 1,3-benzenedicarboxylicacid, 5-amino-, 1,3-bis phenylmethyl ester
7023592
Dibenzyl5-Aminobenzol-1,3-Dicarboxylat
C1=CC=C(C=C1)COC(=O)C2=CC(=CC(=C2)N)C(=O)OCC3=CC=CC=C3
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
152699-63-3
C22H19NO4
361.397
MFCD03093065
VDJWRMQQXQBYIO-UHFFFAOYSA-N
dibenzyl 5-aminoisophthalate, 1,3-dibenzyl 5-aminobenzene-1,3-dicarboxylate, acmc-20alb1, 3,5-dibenzyloxycarbonylaniline, 3,5-dibenzyloxycarbonyl-aniline, 5-amino-isophthalic acid dibenzyl ester, phenylmethyl 5-amino-3-benzyloxycarbonyl benzoate, 1,3-benzenedicarboxylicacid, 5-amino-, 1,3-bis phenylmethyl ester
7023592
Dibenzyl5-Aminobenzol-1,3-Dicarboxylat
C1=CC=C(C=C1)COC(=O)C2=CC(=CC(=C2)N)C(=O)OCC3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Dibenzyl 5-aminoisophthalate
Schmelzpunkt 116°C to 118°C
Menge 1 g
Beilstein 744276
Formelmasse 361.4
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'