missing translation for 'onlineSavingsMsg'
Learn More

Diethyl isopropylidenemalonate, 97%

Product Code. 11367626
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11367626

Brand: Thermo Scientific Alfa Aesar L11195.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 6802-75-1 | C10H16O4 | 200.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6802-75-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00009147
InChI Key WEISAZNMMVPNTH-UHFFFAOYSA-N
Synonym diethyl isopropylidenemalonate, diethyl 2-propan-2-ylidene malonate, diethylisopropylidenemalonate, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester, isopropylidenemalonic acid diethyl ester, 1,3-diethyl 2-propan-2-ylidene propanedioate, diethyl 2-1-methylethylidene malonate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m
PubChem CID 81255
IUPAC Name diethyl 2-propan-2-ylidenepropanedioate
SMILES CCOC(=O)C(=C(C)C)C(=O)OCC

Specifications

Density 1.021
Boiling Point 123°C to 126°C (12 mmHg)
Flash Point 109°C (228°F)
Refractive Index 1.45
Quantity 25 g
Beilstein 1776122
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material Diethyl isopropylidenemalonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.