missing translation for 'onlineSavingsMsg'
Learn More

Difenoconazol-d6, TRC

Artikelnummer. 30382457
Change view
Click to view available options
Menge:
50 mg
Unit Size:
50 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30382457 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382457 Supplier Toronto Research Chemicals Supplier No. TRCD44537250MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Difenoconazole-d6
CAS 2748435-80-3
Hinweise zur Reinheitsqualität HPLC
Summenformel C19H11D6Cl2N3O3
Menge 50 mg
Synonym Difenoconazole D6 (1,1,2,3,3,3-propyl D6), Difenoconazole-d6, Difenoconazole-d6 (1,3-dioxolan-4,5,5-d3-4-methyl-d3) (mixture of diastereomers), Difenoconazole-D6 (4-(methyl-D3)-1,3-dioxolan-2-yl-4,5,5-D3), 1-[[2-[2-Chloro-4-(4-chlorophenoxy)phenyl]-4-(methyl-d3)-1,3-dioxolan-2-yl-4,5,5-d3]methyl]-1H-1,2,4-triazole, Dividend-d6, Dividend (fungicide)-d6, Dragon-d6, Plover-d6, Score-d6, Score 10WG-d6, Score EC 250-d6, Sico-d6, Bardos Neu-d6, CGA 169374-d6, 1-[[2-[2-Chloro-4-(4-chlorophenoxy)phenyl]-4-methyl-1,3-dioxolan-2-yl]methyl]-1H-1,2,4-triazole-d6, Difenoconazole D6 (1,1,2,3,3,3-propyl-D6)
InChI-Formel InChI=1S/C19H17Cl2N3O3/c1-13-9-25-19(27-13,10-24-12-22-11-23-24)17-7-6-16(8-18(17)21)26-15-4-2-14(20)3-5-15/h2-8,11-13H,9-10H2,1H3/i1D3,9D2,13D
SMILES [2H]C([2H])([2H])C1([2H])OC(Cn2cncn2)(OC1([2H])[2H])c3ccc(Oc4ccc(Cl)cc4)cc3Cl
IUPAC-Name 1-[[2-[2-Chlor-4-(4-chlorphenoxy)phenyl]-4,4,5-trideuterio-5-(trideuteriomethyl)-1,3-dioxolan-2-yl]methyl]-1,2,4-triazol
Molekulargewicht (g/mol) 412.3
Formelmasse 411.1
Reinheit (%) >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.