missing translation for 'onlineSavingsMsg'
Learn More

Dihydrexidine hydrochloride, Tocris Bioscience™

Selective D1-like agonist

Marke:  Tocris (Bio-Techne) 08845

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 5mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418113

  • 198.00 € / 5 Milligramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

A potent, full efficacy dopamine D1 agonist which shows no agonist activity at peripheral D2 receptors or adrenoceptors at doses which cause maximal stimulation of D1 sites. The compound appears to be fully bioavailable in brain and exhibits profound antiparkinsonism effects in vivo.
Spezifikation

Chemische Identifikatoren

158704-02-0
303.786
dihydrexidine hydrochloride, dihydrexidine hcl, +/--trans-10,11-dihydroxy-5,6,6a,7,8,12b-hexahydrobenzo a phenanthridine hydrochloride, inverted question mark-trans-10,11-dihydroxy-5,6,6a,7,8,12b-hexahydrobenzo a phenanthridine hydrochloride
(6aR,12bS)-5,6,6a,7,8,12b-hexahydrobenzo[a]phenanthridine-10,11-diol;hydrochloride
C17H18ClNO2
IJYUPBNUPIRQEP-SATBOSKTSA-N
11957519
C1CC2=CC(=C(C=C2C3C1NCC4=CC=CC=C34)O)O.Cl

Spezifikation

Dihydrexidine hydrochloride
158704-02-0
dihydrexidine hydrochloride, dihydrexidine hcl, +/--trans-10,11-dihydroxy-5,6,6a,7,8,12b-hexahydrobenzo a phenanthridine hydrochloride, inverted question mark-trans-10,11-dihydroxy-5,6,6a,7,8,12b-hexahydrobenzo a phenanthridine hydrochloride
C1CC2=CC(=C(C=C2C3C1NCC4=CC=CC=C34)O)O.Cl
303.786
303.79
5mg
C17H18ClNO2
IJYUPBNUPIRQEP-SATBOSKTSA-N
(6aR,12bS)-5,6,6a,7,8,12b-hexahydrobenzo[a]phenanthridine-10,11-diol;hydrochloride
11957519
>99%
Videos