missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogenhexachlorplatinat(IV)-Hexahydrat, niedriges Nitrat, Premion™, 99.95 % (Metallbasis), Pt 37.5 % min, Thermo Scientific Chemicals

Catalog Number p-7058586
Click to view available options
Menge:
1 g
5 g
25 g
18497-13-7
Cl6H14O6Pt
517.89
MFCD00149910
PIJUVEPNGATXOD-UHFFFAOYSA-J
platinum(4+) dihydrogen hexahydrate hexachloride
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
18497-13-7
Cl6H14O6Pt
517.89
MFCD00149910
PIJUVEPNGATXOD-UHFFFAOYSA-J
platinum(4+) dihydrogen hexahydrate hexachloride
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Dihydrogen hexachloroplatinate (IV) hexahydrate
Namenshinweis Low nitrate
Schmelzpunkt 60°C (decomposition)
Farbe Rot bis braun
Dichte 2.430 g/mL
Geruch Odorless
Hinweise zum Prozentgehalt (Metallbasis)
Menge 1 g
UN-Nummer UN2507
Empfindlichkeit Hygroscopic
Merck Index 14,7526
Löslichkeitsinformationen Soluble in water,ethanol,ether,ethyl acetate and acetone. Insoluble in nitric acid.
Formelmasse 517.91 (409.82 Anhydrous)
Reinheit (%) 99.95%
Güte Premion™
Concentration or Composition (by Analyte or Components) PT 37,5 % min
Physikalische Form Klümpchen
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'