Learn More
Dimethylformamide D7 99.50% D, Eurisotop
Product Code. 17267436
Encompass
Change view
Click to view available options
Quantity:
5 mL
2 x 0.75 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17267436 5 mL
17257436 2 x 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17267436 Supplier Eurisotop Supplier No. D110FD
 Restricted Item
Verfügbar ab: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Synonym: N,N-Dimethylformamide-d7, DMF-d7, Heptadeutero-N,N-dimethylformamide
Dimethylformamide D7
  • Isotopic Enrichment: 99.50% D
CoA and MSDS available at Eurisotop.com
Chemical Identifiers
CAS 4472-41-7
Molecular Formula C3H7NO
Molecular Weight (g/mol) 80.14
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
Synonym N,N-Dimethylformamide-d7, DMF-d7, Heptadeutero-N,N-dimethylformamide
IUPAC Name N,N-di(2H3)methyl(2H)formamide
SMILES [2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]
Specifications
Color Colorless
Deuterium (D) 99.50% D
Quantity 5 mL
Melting Point -61°C
Boiling Point 153°C
Grade Research
Chemical Name or Material Dimethylformamide D7