missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride, Tocris Bioscience™

GPR3 agonist; also inhibits NOS and NADPH oxidases

Marke:  Tocris (Bio-Techne) 050410

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13417743

  • 148.50 € / 10 Milligramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

GPR3 agonist (EC50 = 1μM); activates adenylate cyclase through GPR3 but not GPR6 or GPR12. Also induces Ca2+ mobilization and β-arrestin receptor internalization. Binds strongly to flavoproteins; inhibits several enzymes, including NO synthase, NADPH oxidases and NADPH cytochrome P450 oxidoreductase. Also inhibits platelet aggregation.
Spezifikation

Chemische Identifikatoren

4673-26-1
314.55
diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
CHEBI:77967
C12H8ClI
FCFZKAVCDNTYID-UHFFFAOYSA-M
2733504
C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]

Spezifikation

Diphenyleneiodonium chloride
4673-26-1
diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]
2733504
314.55
10mg
C12H8ClI
FCFZKAVCDNTYID-UHFFFAOYSA-M
314.55
CHEBI:77967
>99%
Videos