missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIS3L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25746-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
DIS3L is a probable exonuclease. It has four isoforms.Spezifikation
DIS3L | |
Polyclonal | |
Unconjugated | |
DIS3L | |
DIS3 like exonuclease 1, DIS3L, KIAA1955 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115752 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TF46 | |
DIS3L | |
DIS3L Fusion Protein Ag22689 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |