missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Phenyllaktische Säure, 97 %, Thermo Scientific Chemicals

Catalog Number p-4089528
Click to view available options
Menge:
1 g
10 g
Verpackung:
Glasflasche
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
828-01-3
C9H10O3
166.18
MFCD00065928
VOXXWSYKYCBWHO-UHFFFAOYSA-N
dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
3848
CHEBI:25998
2-Hydroxy-3-phenylpropansäure
C1=CC=C(C=C1)CC(C(=O)O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
828-01-3
C9H10O3
166.18
MFCD00065928
VOXXWSYKYCBWHO-UHFFFAOYSA-N
dl-3-phenyllactic acid, 3-phenyllactic acid, dl-beta-phenyllactic acid, b-phenyllactic acid, 2-hydroxy-3-phenyl-propionic acid, 3-phenyllactate, 2-hydroxy-3-phenylpropionic acid, +--3-phenyllactic acid, dl-phenyllactic acid, beta-phenyllactic acid
3848
CHEBI:25998
2-Hydroxy-3-phenylpropansäure
C1=CC=C(C=C1)CC(C(=O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material DL-3-Phenyllactic acid
Schmelzpunkt 95°C
Farbe Weiß
Infrarotspektrum Echt
Prozentgehaltsbereich 96% min. (HPLC)
Verpackung Glasflasche
Strukturformel C6H5CH2CH(OH)COOH
Menge 1 g
Formelmasse 166.18
Reinheit (%) 97%
Physikalische Form Kristallpulver
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'