missing translation for 'onlineSavingsMsg'
Learn More

DL-Citrullin, 97 %

Catalog Number 15463037
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
627-77-0
C6H13N3O3
175.188
MFCD00007955
RHGKLRLOHDJJDR-UHFFFAOYSA-N
dl-citrulline, citrullin, 2-amino-5-ureidopentanoic acid, dl-2-amino-5-ureidovaleric acid, citrulina, citrulline, dl, ornithine, n5-carbamoyl-, dl, dl-ornithine, n 5-aminocarbonyl, n 5-carbamoyl-dl-ornithine, n 5-aminocarbonyl ornithine
833
CHEBI:18211
2-Amino-5-(carbamoylamino)pentansäure
C(CC(C(=O)O)N)CNC(=O)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
627-77-0
C6H13N3O3
175.188
MFCD00007955
RHGKLRLOHDJJDR-UHFFFAOYSA-N
dl-citrulline, citrullin, 2-amino-5-ureidopentanoic acid, dl-2-amino-5-ureidovaleric acid, citrulina, citrulline, dl, ornithine, n5-carbamoyl-, dl, dl-ornithine, n 5-aminocarbonyl, n 5-carbamoyl-dl-ornithine, n 5-aminocarbonyl ornithine
833
CHEBI:18211
2-Amino-5-(carbamoylamino)pentansäure
C(CC(C(=O)O)N)CNC(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material DL-Citrulline
Menge 25 g
Formelmasse 175.19
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'