missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactic acid, 85+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-152957
Click to view available options
Menge:
500 g
2.5 kg
Verpackung:
EcoSafPak™ Glass Bottle
Glass bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
50-21-5, 7732-18-5
C3H6O3
90.08
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
612
CHEBI:78320
2-hydroxypropanoic acid
CC(O)C(O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
50-21-5, 7732-18-5
C3H6O3
90.08
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
612
CHEBI:78320
2-hydroxypropanoic acid
CC(O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material DL-Lactic acid
Farbe Colorless
Prozentgehaltsbereich 85+%
Strukturformel CH3CH(OH)CO2H
Merck Index 15, 5384
Formelmasse 90.08
Reinheit (%) 85.0 to 90.0%
Güte ACS Reagent
Dichte 1.2000g/mL
Menge 500 g
Verpackung EcoSafPak™ Glass Bottle
Physikalische Form Slightly Viscous Liquid
Glührückstand 0.02% max.
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'