missing translation for 'onlineSavingsMsg'
Learn More

DMSO, sterile filtered, Tocris Bioscience™

Solvent

Marke:  Tocris (Bio-Techne) 3176/100ML

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 100mL

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412564

  • 285.13 € / 100 Milliliter


Voraussichtlicher Liefertermin 26-03-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Solvent with wide ranging applications in biological research. Improves responsiveness of human embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) to differentiation signals.
Spezifikation

Chemische Identifikatoren

67-68-5
78.129
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262
CS(=O)C
C2H6OS
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
methylsulfinylmethane

Spezifikation

DMSO, sterile filtered
67-68-5
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CS(=O)C
78.129
CHEBI:28262
>99.9%
100mL
C2H6OS
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13
Videos