missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH17 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24488-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Spezifikation
DNAH17 | |
Polyclonal | |
Unconjugated | |
DNAH17 | |
DNAH17, DNAHL1, DNEL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8632 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UFH2 | |
DNAH17 | |
DNAH17 Fusion Protein Ag20033 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |