missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
DNAJC10 Polyclonal antibody specifically detects DNAJC10 in Human, Mouse, Rat samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | DNAJC10 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500-1:2000 |
| Zusammensetzung | PBS with 50% glycerol, pH7.3. |
| Gen-Alias | DKFZp434J1813, DnaJ (Hsp40) homolog, subfamily C, member 10, dnaJ homolog subfamily C member 10, ERdj5, ER-resident protein ERdj5, J-domain-containing protein disulfide isomerase-like protein, JPDI, macrothioredoxin, MGC104194, MTHr |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 614-793 of human DNAJC10 (NP_061854.1). IDCQQYHSFCAQENVQRYPEIRFFPPKSNKAYHYHSYNGWNRDAYSLRIWGLGFLPQVSTDLTPQTFSEKVLQGKNHWVIDFYAPWCGPCQNFAPEFELLARMIKGKVKAGKVDCQAYAQTCQKAGIRAYPTVKFYFYERAKRNFQEEQINTRDAKAIAALISEKLETLRNQGKRNKDEL |
| Reinigungsverfahren | Affinity purified |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?