missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dog (Beagle) Cryopreserved Hepatocytes, plateable Female

Dog (beagle) cryopreserved hepatocytes, plateable female

Marke:  Gibco™ DGCP20

Weitere Details : Netto-Gewicht : 0.00280kg

Artikelnummer. 10560861

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »


Beschreibung

Beschreibung

These plateable cryopreserved female dog (Beagle) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.

Cells per vial: 4-8 million

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:
  • Phase I and II enzyme activities
  • Proper morphology
  • Viabilities generally >75%
  • Attachment efficiency
  • Monolayer formation and integrity

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cryopreserved hepatocytes

    Learn about our full line of hepatocytes ,

    Note:
    These products require shipment in LN2 vapor dewars therefore additional fees may apply.
  • Spezifikation

    Spezifikation

    135°C
    Cryopreserved
    Hepatocytes
    Plateable Female Dog (Beagle) Cryopreserved Hepatocytes, 1 x 1.5mL Vial, Dog/Canine Species, Hepatocytes Cell Type, 4-8 x 106 Number of Cells, Single Donor, Liquid Nitrogen Storage, For Hepatocytes and Hepatic Models, Pharma and Biopharma, Primary Cell Culture
    Hepatocytes
    Cell culture, drug discovery and development, primary cell culture, hepatocyte culture, hepatocytes and hepatic models, pharma and biopharma
    1 x 1.5mL Vial
    Produktvorschläge

    Produktvorschläge

    Videos
    Sicherheitsdatenblatt (SDS)
    Dokumentation

    Dokumentation

    Zertifikate
    Sonderangebote

    Sonderangebote

    For Research Use Only. Not for use in diagnostic procedures.