missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15749-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The MW of this protein is 53 kDa, and this antibody specifically recognizes the 53 kDa protein.
DOK3 DOK proteins are enzymatically inert adaptor or scaffolding proteins. They provide a docking platform for the assembly of multimolecular signaling complexes. DOK3 is a negative regulator of JNK signaling in B-cells through interaction with INPP5D/SHIP1. May modulate ABL1 function (By similarity).Spezifikation
DOK3 | |
Polyclonal | |
Unconjugated | |
DOK3 | |
docking protein 3, DOK3, DOKL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27261, 306760, 79930 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B2RYG7, Q7L591, Q9QZK7 | |
Dok3 | |
DOK3 Fusion Protein Ag8221 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |