Learn More
Invitrogen™ Donkey anti-Chicken IgY (H+L) Highly Cross Adsorbed Secondary Antibody, Alexa Fluor™ 555
Beschreibung
This donkey anti-chicken IgY (H+L) whole secondary antibody has been affinity-purified and shows minimum cross-reactivity to bovine, goat, guinea pig, syrian hamster, horse, human, mouse, rabbit, rat and sheep serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondary antibodies flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g. flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 555 dye is a bright, orange-fluorescent dye with excitation ideally suited to the 555 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 555 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 555 dye molecules can be attached to...
Spezifikation
Spezifikation
| Antigen | Chicken IgY (H+L) Highly Cross Adsorbed |
| Anwendungen | Peptide Array, Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konzentration | 2 mg/mL |
| Konjugat | Alexa Fluor 555 |
| Zusammensetzung | PBS with 5mM sodium azide; pH 7.5 |
| Wirtsspezies | Donkey |
| Immunogen | Purified Chicken IgY, Heavy and Light chains. |
| Reinigungsverfahren | Purified |
| Menge | 500 μg |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.