Learn More
Donkey anti-Goat IgG - H&L (Cy5), Secondary Antibody, Abcam™
Marke: Abcam AB6566-500UG
Weitere Details : Netto-Gewicht : 0.25000kg
Beschreibung
Host Species: Donkey
Isotype: IgG
Immunogen: Goat IgG, whole molecule
Applications:
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Spezifikation
Goat IgG - H&L (Cy5) | |
1mg/mL | |
Donkey | |
500 μg | |
Secondary |
Polyclonal | |
Cyanine5 | |
Affinity Purified | |
Immunology | |
IgG |