missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, Texas Red, Invitrogen™
Donkey Polyclonal Secondary Antibody
Marke: Invitrogen PA128662
Weitere Details : Netto-Gewicht : 1.24740kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Goat IgG (H+L) Polyclonal antibody specifically detects Goat IgG (H+L) in Goat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry.Spezifikation
Goat IgG (H+L) | |
Polyclonal | |
Texas Red | |
Donkey | |
IgG fraction | |
RUO | |
Goat | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry | |
2 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
Full length native Goat IgG (purified). | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |