Learn More
Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Qdot™ 655, Invitrogen™
Donkey Polyclonal Secondary Antibody
Marke: Invitrogen Q22089
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Rabbit IgG (H+L) Polyclonal antibody specifically detects Rabbit IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
Rabbit IgG (H+L) | |
Polyclonal | |
Qdot 655 | |
Donkey | |
purified | |
RUO | |
Rabbit | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 μM | |
0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
Gamma Immunoglobins Heavy and Light chains | |
100 μL | |
Secondary | |
4° C, store in dark | |
IgG |