Learn More
Dräger™ Bromine (Cl2) XXS Replacement Sensor
Description
- Measuring range: 0 to 20 ppm
- Resolution: 0.1 ppm
- Bromine (Cl2)
Specifications
Specifications
| Components Measured | Bromine |
| For Use With (Equipment) | Pac 7000, X-am 5000 Multigas Detectors |
| Type | Replacement Sensor |