Learn More
Draeger DraegerSensor XXS Miniature Electrochemical Sensor
Description
- For portable DraegerSensor gas detection devices
- Reliably and quickly alerts users to the presence of hazardous gases and vapours
- Low operating costs for longer use
Specifications
Specifications
| Description | Miniature electrochemical sensor |
| For Use With (Equipment) | Draeger Pac, X-am 1/2/5 and X-am 8000 Generation Detectors |
| Type | Multiple Gas Detector Sensor |
| For Use With (Application) | Multiple Hazardous Gas and Vapour Detection |