Beschreibung
DRAP1 Monoclonal antibody specifically detects DRAP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | DRAP1 |
| Anwendungen | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | PCRP-DRAP1-1A8 |
| Konzentration | 0.2 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Zusammensetzung | 10 mM PBS with 0.05% BSA |
| Gen-Alias | dr1-associated corepressor, Dr1-associated protein 1, DR1-associated protein 1 (negative cofactor 2 alpha), NC2-alphaNegative co-factor 2-alpha, negative cofactor 2 alpha |
| Wirtsspezies | Mouse |
| Mehr anzeigen |