missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DUSP26 Polyclonal specifically detects DUSP26 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | DUSP26 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS, 2% Sucrose |
| Gen-Alias | DSP-4, dual specificity phosphatase 26 (putative), Dual specificity phosphatase SKRP3, dual specificity protein phosphatase 26, dual-specificity phosphatase SKRP3, DUSP24SKRP3, EC 3.1.3.16, EC 3.1.3.48, LDP4, LDP-4, Low-molecular-mass dual-specificity phosphatase 4, MAP kinase phosphatase 8, MGC1136, MGC2627, Mitogen-activated protein kinase phosphatase 8, MKP8, MKP-8, NATA1, Novel amplified gene in thyroid anaplastic cancer |
| Wirtsspezies | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of mouse DUSP26 |
| Reinigungsverfahren | Immunogen affinity purified |
| Menge | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?