Learn More
Brookfield™ DV3T Rheometer
Product Code. 15822197
Change view
Click to view available options
Interface:
Digital touchscreen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Interface
15822197 Digital touchscreen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15822197 Supplier Brookfield™ Supplier No. XDX3TRVKB00G00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Quick, straightforward, single point viscosity is easily accomplished with the Brookfield™ DV3T rheometer, making it the ideal all-in-one instrument for busy QC labs as well as demanding R&D departments doing comprehensive flow behaviour. Spindles are included.
  • The programmable DV3T offers the most advanced user experience in viscosity testing and measurement
  • Full color 7 in. (18 cm) touchscreen conveniently displays all test parameters along with measured values for yield stress, viscosity and temperature
  • On screen graphing
  • Built-in models can provide rapid data analysis for flow index and other selectable variables
  • PG Flash software allows the operator to create repeatable custom tests on a PC using the same controls they see on the DV3T for upload to the unit (software and flash drive are included)
  • Enhanced security elements such as customisable user levels and password access
Specifications
Description Rheometer with spindles (6) included
Interface Digital touchscreen
Measuring Accuracy ±1.0% of range
Rotational Speed Range 0.01 to 250 rpm
Temperature Range (Metric) 0°C to 40°C
Viscosity Range 100 cP to 40 McP