missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYX1C1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14522-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a tetratricopeptide repeat domain-containing protein. The encoded protein interacts with estrogen receptors and the heat shock proteins, Hsp70 and Hsp90. A chromosomal translocation involving this gene is associated with a susceptibility to developmental dyslexia. Mutations in this gene are associated with deficits in reading and spelling. Alternate splicing results in multiple transcript variants.Spezifikation
DYX1C1 | |
Polyclonal | |
Unconjugated | |
DNAAF4 | |
DYX1, DYX1C1, DYXC1, EKN1, RD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
161582, 363096, 67685 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5VJS5, Q8R368, Q8WXU2 | |
Dnaaf4 | |
DYX1C1 Fusion Protein Ag5997 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |